Caffeine, Calbiochem®

Supplier: MilliporeSigma
Warning

205548-500GM
80055-528EA 164.61 USD
80055-528
Caffeine, Calbiochem®
Caffeine

Trimethylated xanthine that acts as a potent central nervous system stimulant. At higher doses, produces tachycardia and interferes with the uptake and storage of Ca2+ by sarcoplasmic reticulum in skeletal muscle. Prevents apoptosis and cell cycle effects induced by camptothecin (80055-998) and topotecan in HL-60 cells. Heavy metals: <=10ppm.

Formula: C₈H₁₀N₄O₂
Melting Pt: 235…239 °C
Density: 1.23 g/cm³ (19 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005758
CAS Number: 58-08-2
EINECS: 200-362-1
Merck Index: 13,01636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07e609e1c7a199176b9e5b938ef241d166d502f9&vid=1ac1145bba3574e2401439da90f37fee7fd7610b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR