Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3)

Supplier: MilliporeSigma

Synonyms: Uridine-5'-triphosphoric acid trisodium salt, Uridine triphosphate trisodium salt

6701-50MG
80055-488EA 92.94 USD
80055-488
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3)
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3)
Formula: C₉H₁₂N₂Na₃O₁₅P₃
MW: 550.09 g/mol
MDL Number: MFCD00044310
CAS Number: 19817-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR