Toluene-α-sulfonyl fluoride ≥98%, Calbiochem®

Supplier: MilliporeSigma
Danger

52332-25GM 52332-5GM 52332-1GM
80055-382EA 415.56 USD
80055-382 80055-384 80055-380
Toluene-α-sulfonyl fluoride ≥98%, Calbiochem®
Toluene-α-sulphonyl fluoride

Irreversible inhibitor of serine proteases. Its mechanism of action is analogous to that of diisopropylfluorophosphate. PMSF causes sulfonylation of the active-site serine residues. Also reported to inhibit internucleosomal DNA fragmentation in immature thymocytes. Effective concentration: 50µM.

Formula: C₇H₇FO₂S
Boiling Pt: 80…85 °C (0.3 mmHg)
Melting Pt: 91…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00007424
CAS Number: 329-98-6
EINECS: 206-350-2
UN: 2923
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR