Lipopolysaccharides (from S. typhimurium)
Supplier: MilliporeSigma
437650-5MG
80055-316EA 290.85 USD
80055-316
Lipopolysaccharides (from S. typhimurium)
Lipopolysaccharides
Stimulates the activity of inducible nitric oxide synthase
Induces apoptosis in mouse thymus and swine lymphocytes.