Diptheria Toxin, MilliporeSigma
Supplier: MilliporeSigma
322326-1MG
80055-232EA 1459.19 USD
80055-232
Diptheria Toxin, MilliporeSigma
Microbiology Test Systems Antigen-Antibody Tests Ag-AK, Seren
Diphtheria toxin Catalyzes ADP-ribosylation of eukaryotic aminoacyltransferase II (EF2) using NAD as substrate, thereby inhibiting protein synthesis
  • Induce internucleosomal breakdown
  • It can cause DNA fragmentation and cytolysis in U937 cells
  • Activation requires nicking with a protease followed by reduction with DTT
  • Storage: +2 °C...+8 °C
For assays employing very low concentrations of diphtheria toxin, the use of a carrier protein, such as BSA or HSA, is recommended.The primary target is Eukaryotic aminoacyltransferase II (EF2)
This toxin is from Corynebacterium diphtheriae. This toxin is highly harmful