Cholera Toxin, Vibrio cholerae, Type Inaba 569B, Azide Free, MilliporeSigma
Supplier: MilliporeSigma
227036-1MG
80055-160EA
451.62
USD
80055-160
Cholera Toxin, Vibrio cholerae, Type Inaba 569B, Azide Free, MilliporeSigma
Microbiology Test Systems
Antigen-Antibody Tests
Ag-AK, Seren
Cholera toxin has a single A subunit surrounded by five polypeptides chains of the B subunits
Useful for studies involving adenylate cyclase, AMP, membrane-transport phenomena and for tissue culture applications. The A subunit catalyzes the ADP-ribosylation of the α-subunit of G-proteins, reducing intrinsic GTPase activity and activating the α-subunit. The B subunits are responsible for the attachment of the native toxin on the cell surface.