Caspase-1 Substrate VI, Fluorogenic, Calbiochem®
Supplier: MilliporeSigma
688225-5MG
80055-026EA 507.2 USD
80055-026
Caspase-1 Substrate VI, Fluorogenic, Calbiochem®
Electrophoresis Reagents Western Blotting Reagents
Fluorogenic caspase-1 substrate
Reaction can be monitored visually or quantitatively by a blue to green shift in fluorescence upon cleavage of the AFC fluorophore. Excitation maximum: ~400nm; emission maximum: ~505nm.