Uridine-5′-diphosphogalactose disodium salt

Supplier: MilliporeSigma

670111-50MG 670111-10MG
80054-962EA 185.58 USD
80054-962 80054-960
Uridine-5′-diphosphogalactose disodium salt
Uridine-5′-diphosphogalactose disodium salt
Formula: C₁₅H₂₃N₂Na₂O₁₇P₂
MDL Number: MFCD00077895
CAS Number: 137868-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR