Tetrahydrouridine, Calbiochem®

Supplier: MilliporeSigma

Synonyms: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-4-hydroxytetrahydropyrimidin-2(1H)-one

584222-10MG
80054-664EA 292.98 USD
80054-664
Tetrahydrouridine, Calbiochem®
Tetrahydrouridine

Tetrahydrouridine is a potent, competitive inhibitor of cytidine deaminase.


  • Competitive inhibitor of cytidine deaminase
  • Assess anti-leukemic activitiy of Ara-C
  • Used in in vitro studies


A potent competitive inhibitor of cytidine deaminase. Used in combination with cytosine arabinoside (Ara-C) to assess the anti-leukemic activity and anti-tumor activity of Ara-C in in vitro studies.

Formula: C₉H₁₆N₂O₆
MW: 248.2331 g/mol
MDL Number: MFCD00063468
CAS Number: 18771-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR