Pluronic® F-127 10% in aqueous solution, PROTEIN GRADE® detergent, sterile-filtered, Calbiochem®

Supplier: MilliporeSigma

540025-50ML
80054-134EA 376.5 USD
80054-134
Pluronic® F-127 10% in aqueous solution, PROTEIN GRADE® detergent, sterile-filtered, Calbiochem®
Poloxamer 407 (Pluronic® F-127)

A non-ionic detergent useful for isolation of membrane proteins. It has been used to solubilize large dye molecules in physiological media. Because of its low UV absorbance, it can be used in protocols that require UV monitoring of the solubilized proteins.

Formula: HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)aH
Storage Temperature: Freezer
MDL Number: MFCD00082049
CAS Number: 9003-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR