Proteasome Substrate III, Fluorogenic, Calbiochem®
Supplier: MilliporeSigma
539142-5MG
80053-860EA 394.17 USD
80053-860
Proteasome Substrate III, Fluorogenic, Calbiochem®
Electrophoresis Reagents Western Blotting Reagents
Fluorogenic proteasome substrate that also acts as a substrate for calpain, chymotrypsin, and ingensin.
Excitation maximum: ~380nm. Emission maximum: ~460nm.