Proteasome Substrate II, Fluorogenic, Calbiochem®
Supplier: MilliporeSigma
539141-5MG
80053-858EA 285.6 USD
80053-858
Proteasome Substrate II, Fluorogenic, Calbiochem®
Electrophoresis Reagents Western Blotting Reagents
Fluorogenic proteasome substrate.
Excitation maximum: ~380nm. Emission maximum: ~460nm.