Lecithin (from egg yolk)

Supplier: MilliporeSigma

524617-250MG
80053-676EA 85.9 USD
80053-676
Lecithin (from egg yolk)
Lecithin

A lipotropic agent, chromatographically purified.


  • Synonym: L-α-Phosphatidylcholine

MDL Number: MFCD00147406
CAS Number: 8002-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR