Mouse Recombinant Nitric Oxide Synthase, EMD Millipore
Supplier: MilliporeSigma
482777-50U
80053-412EA 591.96 USD
80053-412
Mouse Recombinant Nitric Oxide Synthase, EMD Millipore
Enzymes
In 25 mM HEPES, containing 8 µM tetrahydro-L-biopterin, 10% glycerol, pH 7.4. Specific activity: >=7 units/mg protein.