MnTBAP
Supplier: MilliporeSigma
475870-25MG
80053-226EA 114.99 USD
80053-226
MnTBAP
Enzymes
Cell-permeable superoxide dismutase (SOD) mimetic and peroxynitrite scavenger. Protects endothelial cells in a dose-dependent manner (EC50~40 µM) against paraquat (2 mM). Inhibits the oxidation of Dihydrorhodamine 123 by peroxynitrite, but does not scavenge nitric oxide (NO).