Calbiochem® Human IgE, Myeloma, Immunoassay Standard, EMD Millipore
Supplier: MilliporeSigma
Human IgE, Myeloma is validated for use as an immunoassay standard.
- Formulation: In 150 mM NaCl, 15 mM potassium phosphate buffer, pH 7.4
- Preservative: ≤0.1% sodium azide
- Purity: Single arc by immunoelectrophoresis against antiserum to whole human serum and human IgE
- Source: Prepared from plasma that was shown by certified tests to be nonreactive for HBsAg and for antibodies to HCV, HBc, HIV-1 and HIV-2
The heavy chain of myeloma IgE may appear as either a single or double band on gel electrophoresis. Variables associated with assay conditions will dictate the proper working dilution.
Packaging: Packed in plastic ampoule.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...