Guanosine-3',5'-cyclicmonophosphoric acid sodium salt

Supplier: MilliporeSigma

Synonyms: Sodium (4aR,6R,7R,7aS)-6-(2-amino-6-oxo-1,6-dihydro-9H-purin-9-yl)-7-hydroxytetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-2-olate 2-oxide, Cyclic GMP sodium

370656-50MG
80051-960EA 123.96 USD
80051-960
Guanosine-3',5'-cyclicmonophosphoric acid sodium salt
Guanosine-3',5'-cyclicmonophosphoric acid sodium salt
Formula: C₁₀H₁₁N₅NaO₇P
MW: 367.19 g/mol
MDL Number: MFCD00038424
CAS Number: 40732-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR