Granzyme B Substrate I, Colorimetric, Calbiochem®, MilliporeSigma
Supplier: MilliporeSigma
368057-5MG
80051-940EA 274.94 USD
80051-940
Granzyme B Substrate I, Colorimetric, Calbiochem®, MilliporeSigma
Electrophoresis Reagents Western Blotting Reagents
Colorimetric substrate for the detection of granzyme B and caspase-8 activity. Cleavage of pNA is monitored colorimetrically at ~405nm.