Genistein, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
5,7-Dihydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one, 4',6'-Dihydroxy-3-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',5,7-Trihydroxyisoflavone
Inhibits protein tyrosine kinases, including autophosphorylation of epidermal growth factor receptor kinase (IC50=2.6µM)
The inhibition is competitive with respect to ATP and non-competitive with respect to the phosphate acceptor. Has only a trivial effect on the activity of PKA and PKC (IC50>350µM). Inhibits tumor cell proliferation and induces tumor cell differentiation. Produces cell cycle arrest and apoptosis in Jurkat T-leukemia cells. However, it prevents anti-CD3 monoclonal antibody-induced thymic apoptosis. Also inhibits topoisomerase II activity in vitro.
Formula:
C₁₅H₁₀O₅ |
MDL Number:
MFCD00016952 CAS Number: 446-72-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...