BAPTA tetrasodium salt, Calbiochem®, Millipore®

Supplier: MilliporeSigma

196418-1GM
80050-386EA 313.83 USD
80050-386
BAPTA tetrasodium salt, Calbiochem®, Millipore®
BAPTA tetrasodium salt

Ca2+ chelator exhibiting a 105-fold greater affinity for Ca2+ .

Formula: C₂₂H₂₄N₂O₁₀
Storage Temperature: Freezer
MDL Number: MFCD00067691
CAS Number: 126824-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR