Perfect Protein™ Markers, MilliporeSigma
Supplier: MilliporeSigma
69079-3
80030-960EA 103.22 USD
80030-960 80030-952
Perfect Protein™ Markers, MilliporeSigma
Electrophoresis Reagents Molecular Weight Markers
Ideal for routine use in SDS-polyacrylamide gel electrophoresis.
  • Ready to use
  • Produce sharp, accurately sized bands
These markers contain no oligosaccharides that cause anomalous migration, heterogeneous fuzzy bands, or inaccurate size estimation. The markers are optimized for use with Coomassie blue staining, but adjusted amounts can also be used with other gel staining methods such as silver staining and fluorescent dyes.