Perfect DNA™ Ladder, MilliporeSigma
Supplier: MilliporeSigma
Evenly spaced DNA markers in ready-to-load format.
- Ready to load
- Ideal for routine use
The Perfect DNA™ ladders contain sets of DNA species having convenient, easy-to-remember sizes for agarose gel analysis. The markers have uniform band intensities and cover a wide range of DNA sizes.
Delivery information: An extra vial of 6X Loading Buffer or 6X DNA Gel Loading Buffer is included.