ProteoExtract® Transmembrane Protein Extraction Kit, MilliporeSigma
Supplier: MilliporeSigma
71772-3
80017-516EA 669.59 USD
80017-516
ProteoExtract® Transmembrane Protein Extraction Kit, MilliporeSigma
Protein Extraction Protein Extraction Kits
ProteoExtract® Transmembrane Protein Extraction Kit (TM-PEK) uses a novel, detergent-free chemistry for
Each TM-PEK kit provides sufficient reagents to process a total of 40 samples (20 samples using Extraction Buffer 2A at the recommended dilution, and 20 samples using Extraction Buffer 2B at the recommended dilution). Each sample may be comprised of 1–5 x 107 cultured cells or 25–50 mg of tissue.
Delivery information: Kit includes extraction Buffer 1 (40 mL), extraction Buffer 2 (50 mL), TM-PEK Reagent A (2 mL), TM-PEK Reagent B (2 mL), Protease Inhibitor Cocktail Set III (0.4 mL).