10-NCP 95 HPLC_ASSAY_METHOD, Calbiochem®

Supplier: MilliporeSigma

124020-5MG
80017-188EA 433.89 USD
80017-188
10-NCP 95 HPLC_ASSAY_METHOD, Calbiochem®
10-NCP

10-(4'-(N-diethylamino)butyl)-2-chlorophenoxazine, HCl


White solid. This cell-permeable, selective inhibitor of the phosphorylation of Akt and its in vitro kinase activity has a minimal effect on PI 3-K, PDK1, or SGK1. It is shown to suppress growth of rhabdomyosarcoma cell lines, inhibit IGF-I-stimulated nuclear translocation of Akt, and prevent phosphorylation of the downstream targets mTOR, p70S6 kinase, and S6 ribosomal protein. The mode of inhibition is not PH domain-dependent.


Packaged under inert gas. Hygroscopic. Protect from light. Store at 4°C (39°F). Following reconstitution, aliquot and freeze at –20°C (–4°F). Stock solutions are stable for up to three months when frozen. For research use only, not intended for human or diagnostic use.

Formula: C₂₀H₂₅ClN₂O·HCl
MW: 381.34 g/mol
MDL Number: MFCD09038570
CAS Number: 925681-41-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR