Paragon Scientific Color Standard USP (631) Ferric Chloride CS
Supplier: LGC Standards
ALK-USPPRYE
77976-032EA
158
USD
77976-032
Paragon Scientific Color Standard USP (631) Ferric Chloride CS
Standards
Color Reference and Spectroscopy Standards
Matrix: Aq. HCl
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...