Paragon Scientific Color Standard USP (631) Cobaltous Chloride CS
Supplier: LGC Standards
ALK-USPPRRE
77976-031EA
235
USD
77976-031
Paragon Scientific Color Standard USP (631) Cobaltous Chloride CS
Standards
Color Reference and Spectroscopy Standards
Matrix: Aq. HCl
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...