Paragon Scientific Relative Density 25 °C (Nominal: 0.8352 @ 25 °C)
Supplier: LGC Standards
ALK-RDEN25-04
77975-959EA
483
USD
77975-959
Paragon Scientific Relative Density 25 °C (Nominal: 0.8352 @ 25 °C)
Standards
Density Standards
- Shipping conditions: Room temperature
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...