Paragon Scientific Relative Density 25 °C (Nominal: 0.7101 @ 25 °C)
Supplier: LGC Standards
ALK-RDEN25-02
77975-957EA
483
USD
77975-957
Paragon Scientific Relative Density 25 °C (Nominal: 0.7101 @ 25 °C)
Standards
Density Standards
- Shipping conditions: Room temperature
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...