Paragon Scientific Relative Density 15 °C (Nominal: 0.8494 @ 15 °C)
Supplier: LGC Standards
ALK-RDEN15-05
77975-947EA
483
USD
77975-947
Paragon Scientific Relative Density 15 °C (Nominal: 0.8494 @ 15 °C)
Standards
Density Standards
- Shipping conditions: Room temperature
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...