Paragon Scientific Refractive Index (Nominal: 1.5440 @ 25 °C), Certified @ 20, 25 and 30 °C
Supplier: LGC Standards
ALK-PS-RI-09K
77975-913EA
0
USD
77975-913
Paragon Scientific Refractive Index (Nominal: 1.5440 @ 25 °C), Certified @ 20, 25 and 30 °C
Standards
Refractive Index Standards
Standard type: Single component
Shipping conditions: Room temperature
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...