Paragon Scientific Refractive Index (Nominal: 1.3325 @ 25 °C), Certified @ 20, 25 and 30 °C
Supplier: LGC Standards
ALK-PS-RI-01K
77975-904EA 0 USD
77975-904
Paragon Scientific Refractive Index (Nominal: 1.3325 @ 25 °C), Certified @ 20, 25 and 30 °C
Standards Refractive Index Standards
  • Standard type: Single component
  • Shipping conditions: Room temperature