Paragon Scientific Density 50 °C (Nominal: 0.7864 g/ml @ 50 °C)
Supplier: LGC Standards
ALK-DEN50-01
ALK-DEN50-01-500ML
77975-712EA
97
USD
77975-712
77975-713
Paragon Scientific Density 50 °C (Nominal: 0.7864 g/ml @ 50 °C)
Standards
Density Standards
- Shipping conditions: Room temperature
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...