Paragon Scientific Density 20 °C (Nominal: 1.1915 g/ml @ 20 °C)
Supplier: LGC Standards
ALK-DEN20-12-500ML
77975-678EA
570
USD
77975-678
Paragon Scientific Density 20 °C (Nominal: 1.1915 g/ml @ 20 °C)
Standards
Density Standards
- Shipping conditions: Room temperature
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...