Paragon Scientific Density 20 °C (Nominal: 1.1256 g/ml @ 20 °C)
Supplier: LGC Standards
ALK-DEN20-11-500ML
77975-677EA
570
USD
77975-677
Paragon Scientific Density 20 °C (Nominal: 1.1256 g/ml @ 20 °C)
Standards
Density Standards
- Shipping conditions: Room temperature
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...