Paragon Scientific Density 15 °C (Nominal: 0.9413 g/ml @ 15 °C)
Supplier: LGC Standards
ALK-DEN15-08-500ML
77975-651EA
570
USD
77975-651
Paragon Scientific Density 15 °C (Nominal: 0.9413 g/ml @ 15 °C)
Standards
Density Standards
- Shipping conditions: Room temperature
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...