Paragon Scientific Density 15 °C (Nominal: 0.6654 g/ml @ 15 °C)
Supplier: LGC Standards
ALK-DEN15-01
ALK-DEN15-01-500ML
77975-640EA
97
USD
77975-640
77975-641
Paragon Scientific Density 15 °C (Nominal: 0.6654 g/ml @ 15 °C)
Standards
Density Standards
- Shipping conditions: Room temperature
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...