Paragon Scientific Density 150 °C (Nominal: 0.8287 g/ml @ 150 °C)
Supplier: LGC Standards
ALK-DEN150-03
ALK-DEN150-03-500ML
77975-638EA
97
USD
77975-638
77975-639
Paragon Scientific Density 150 °C (Nominal: 0.8287 g/ml @ 150 °C)
Standards
Density Standards
- Shipping conditions: Room temperature
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...