Paragon Scientific Density 150 °C (Nominal: 0.7816 g/ml @ 150 °C)
Supplier: LGC Standards
ALK-DEN150-02
ALK-DEN150-02-500ML
77975-636EA
97
USD
77975-636
77975-637
Paragon Scientific Density 150 °C (Nominal: 0.7816 g/ml @ 150 °C)
Standards
Density Standards
- Shipping conditions: Room temperature
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...