Paragon Scientific Density 100 °C (Nominal: 0.7645 g/ml @ 100 °C)
Supplier: LGC Standards
ALK-DEN100-01
ALK-DEN100-01-500ML
77975-628EA
97
USD
77975-628
77975-629
Paragon Scientific Density 100 °C (Nominal: 0.7645 g/ml @ 100 °C)
Standards
Density Standards
- Shipping conditions: Room temperature
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...