Paragon Scientific Density Standard, Biodiesel
Supplier: LGC Standards
ALK-CRMU-DEB100
77975-564EA
251
USD
77975-564
Paragon Scientific Density Standard, Biodiesel
Standards
Density Standards
- Shipping conditions: Room temperature
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...