Paragon Scientific 500000 µS/cm @ 25 °C, Conductivity (NIST Traceable, ACS)
Supplier: LGC Standards
ALK-COND500K-25-500ML
77975-553EA 127 USD
77975-553
Paragon Scientific 500000 µS/cm @ 25 °C, Conductivity (NIST Traceable, ACS)
Standards Conductivity Standards