BalanCD™ CHO DG44
Supplier: FUJIFILM IRVINE SCIENTIFIC INC.
91177-1L 94148-10L
77975-345EA 170.75 USD
77975-345 77975-399
BalanCD™ CHO DG44
Cell Culture Media
BalanCD™ CHO DG44 is an animal component-free, chemically defined medium that is specifically designed for optimized growth and productivity of high density suspension cultures. The medium delivers consistent productivity from CHO DG44 cultures for cost-effective biomolecule drug development and commercialization. Support high cell density cultures and achieve robust titers of monoclonal antibodies (mAbs) and recombinant proteins with chemically defined BalanCD™ CHO DG44. The medium delivers consistent cell productivity in suspension cultures for cost-effective biomolecule drug development and commercialization.
  • Achieves high titers of monoclonal antibodies and recombinant proteins for commercial drug development
  • Versatile and scalable for small-scale shake flask and small-to-large bioreactor scale
  • Supports a variety of DG44 cell strains in both batch and fed-batch cultures
  • Pair with BalanCD CHO Feed 4 for optimal mAbs production in fed-batch cultures
  • Scalable and adaptable to use with a variety of CHO DG44 cell strains in both batch and fed-batch cultures
  • Efficient, cost-effective workflow solution for CHO DG44 cells for small- to large-scale bioreactors
  • Compatible with a range of culture platforms and transfection reagents
BalanCD CHO DG44 is a chemically defined growth medium designed specifically for the growth and productivity of suspension CHO DG44 cell lines. BalanCD CHO DG44 is provided without L-Glutamine to extend shelf life. The recommended L-glutamine concentration to add is 4 to 8 mM. Glucose should be monitored during culture as additional supplementation may be required. The medium does not contain antibiotics, antimycotics, protein hydrolysates or any other undefined components.
Packaging:
Store at 2 to 8 °C, protect from light
Shelf life: 12 months (liquid) or 24 months (powder) from date of manufacture