BalanCD™ CHO Feed 2
Supplier: FUJIFILM IRVINE SCIENTIFIC INC.
91129-1L 94121-10L
77975-322EA 202.63 USD
77975-322 77975-394
BalanCD™ CHO Feed 2
Cell Culture Supplements and Additives
BalanCD CHO Feed 2 is a chemically-defined, animal component-free feed medium designed to increase process yields of antibodies and recombinant proteins in Chinese Hamster Ovary (CHO) cells in fed-batch mode. These feeds have the capability to more than double the growth and protein production when compared to a batch culture. BalanCD CHO Feed 1 was designed to be used in conjunction with BalanCD CHO Growth A. BalanCD CHO Feed 2 was designed to be used with a wide range of growth media for optimal performance in fed-batch cultures.
  • All BalanCD CHO Growth Media and Feeds are formulated for industrial applications and transition smoothly into commercial production
  • Purpose-built formulations for fed batch bioprocesses
  • Rational Culture Media Design approach ensures scalability to commercial production
  • BalanCD CHO Feeds are formulated using chemically-defined components in high concentrations based on typical CHO cell requirements
The BalanCD™ CHO Feed 2 has been tested with three model cell lines (CHO-S, CHO-M, and CHO DHFR). BalanCD™ CHO Feed 2 is comprised solely of animal component-free, chemically defined components, contains no antibiotics or antimycotics, and has proven stability in both powder and liquid formats.
Packaging: Store at 2 to 8 °C. Shelf life: 1 year from date of manufacture.