EDTA (Ethylenediamine tetraacetic acid) solution 0.526 mM (Versene 1:5000)

Supplier: FUJIFILM IRVINE SCIENTIFIC INC.

9314-100ML
77975-380EA 24.75 USD
77975-380
EDTA (Ethylenediamine tetraacetic acid) solution 0.526 mM (Versene 1:5000)
EDTA (Ethylenediamine tetraacetic acid)

EDTA (Versene) Solution 0.526 mM (1:5000) is a chelating agent used to life some cell types from the culture vessel surface into suspension.



    • 200 mg EDTA tetrasodium salt per liter
    • Phosphate Buffered Saline solution


Without: Calcium and Magnesium salts


Formula: C₁₀H₁₆N₂O₈
MDL Number: MFCD00003541
CAS Number: 60-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR