HBSS 10X-Hanks' Balanced Salt Solution
Supplier: FUJIFILM IRVINE SCIENTIFIC INC.
9230-500ML
77975-368EA 59.63 USD
77975-368
HBSS 10X-Hanks' Balanced Salt Solution
Buffers Cell Culture Buffers
HBSS 10X Hanks' Balanced salt solution is designed to maintain pH and osmotic balance and to provide cells with water and essential inorganic ions. HBSS is suitable for use in sealed culture flasks incubated in air.
  • Phenol red
Packaging:
Store at 15 to 30 °C and protect from light
Shelf life: 24 months from date of manufacture