BalanCD™ HEK293 Feed
Supplier: FUJIFILM IRVINE SCIENTIFIC INC.
91166-500ML
77975-341EA 166.63 USD
77975-341
BalanCD™ HEK293 Feed
Cell Culture Supplements and Additives
BalanCD™ HEK293 Feed is formulated to significantly increase protein yields in suspension-adapted HEK293 cell lines post-transfection.
  • Increase viable cell density and culture duration to maximize yield
  • Use the same Feed for a wide range of HEK293 cell lines
  • Fulfill regulatory requirements
  • Chemically-defined, animal component-free formula
  • Drug Master File (DMF) submitted to US FDA
The BalanCD HEK293 System is a chemically defined, animal component-free platform of media and supplement optimized for production of viral vectors and transient protein expression. The product system comprises of BalanCD HEK293 medium, BalanCD HEK293 Feed, and Anti-Clumping Supplement. BalanCD HEK293 System contains no hydrolysates, L-Glutamine, antibiotics, antimycotics, or any other undefined components, and is ready to use for suspension culture applications. BalanCD HEK293 medium contains 6 g/L glucose and BalanCD HEK293 Feed contains 40 g/L glucose. BalanCD HEK293 medium can be supplemented with BalanCD HEK293 Feed for high density cell culture, or with Anti-Clumping Supplement post-transfection to minimize cell aggregation.
Packaging:
Store at 2 to 8 °C
Shelf life: One year from date of manufacture