BalanCD™ Transfectory Supplement
Supplier: FUJIFILM IRVINE SCIENTIFIC INC.
91148-500ML
77975-334EA 113.15 USD
77975-334
BalanCD™ Transfectory Supplement
Cell Culture Supplements and Additives
Part of the BalanCD™ Transfectory CHO system of media and supplements, the Transfectory supplement is an animal-component-free concentrated hydrolysate supplement that can be used to enhance cell growth and protein expression in CHO and various cell types.
  • Hydrolysate supplement that can be used to enhance cell growth and protein expression in CHO and various cell types
Transfectory Supplement is concentrated hydrolysate supplement designed to enhance cell growth and protein expression of CHO and various cell types. This supplement is free of animal-derived components.
Packaging:
Store at 2 to 8 °C
Shelf life: One year from date of manufacture