PRIME-XV IS21 Supplement (50X)
Supplier: FUJIFILM IRVINE SCIENTIFIC INC.
91142-10ML
77975-332EA 127.25 USD
77975-332
PRIME-XV IS21 Supplement (50X)
Cell Culture Supplements and Additives
Neural supplement for long-term neuronal cultures.
  • Neural supplement for use with PRIME-XV Neural Basal
  • Medium and conventional media such as Ham’s DMEM/F12 with L-glutamine
  • Does not contain antibiotics
PRIME-XV IS21 Supplement (50X) is a serum-free supplement optimized for use with PRIME-XV Neural Basal Medium to support maintenance and long-term culture of neuronal cells. This supplement is supplied as a 50X concentrated solution and should be diluted to 1X in basal medium before use. This supplement does not contain antibiotics. Designed and manufactured to facilitate transfer from research to clinic. Supported by robust raw material controls and supply chain management.
Certifications: FDA, Federal, and State registered - cGMP-compliant manufacture; EN ISO 13485:2016 certified; MDSAP certified; Extensive QC testing including functionality, sterility (USP <71>), and endotoxin (USP <85>); Drug Master Files (DMFs) filed with the FDA – available upon request.