Metal Tubings, Vacuum Insulated Stainless Steel
Supplier: JULABO
8891818 8891801 8891802 8891803 8891804 8891805 8891806 8891807 8891808 8891809 8891810 8891811 8891812 8891813 8891814 8891815 8891816 8891817
77975-254EA 4076 USD
77975-254 77975-220 77975-222 77975-224 77975-226 77975-228 77975-230 77975-232 77975-234 77975-236 77975-238 77975-240 77975-242 77975-244 77975-246 77975-248 77975-250 77975-252
Metal Tubings, Vacuum Insulated Stainless Steel
Tubing Vacuum/Pressure Tubing
Stainless steel vacuum insulated tubing (–100 to +400 °C) with M16×1, M24×1.5, M30×1.5 and M38×1.5 female fittings. For use with circulators, heaters, chillers, temperature control units with M threaded pump connections.
  • Engineered for cryogenic to high-temperature applications
  • Cleanroom-compatible materials (316L SS)
  • Low spring back reduces stress on connections and joints
  • Improves thermal energy efficiency
  • Maintains a safer external surface temperature
  • Minimizes heat loss and conserves thermal energy
  • Reduces condensation and moisture leaks
Vacuum-insulated metal tubing is the ideal accessory for high and low-temperature applications, including those powered by JULABO PRESTO systems. The insulated tubing prevents frost build-up and maintains optimal cooling capacity.
This type of tubing also increases energy efficiency, decreases heat loss, and provides better temperature accuracy for faster processing. The flexible tubing makes connecting the application to the temperature control unit easy and hassle-free.
Temperature range: –100 to +400 °C
Ordering information:
One-year warranty from ship date.
Delivery information:
Includes lock-tite thread lubricant.
Packaging:
Sold as each.
Caution:
For optimal flow, install the tubing as straight as possible. Do NOT use Teflon tape on M theaded fittings.