Counstar Spica M1 Real-Time Live Cell Imaging and Analysis System
Supplier: ALIT LIFETECH INC
P020400021-A
77973-670EA 39999 USD
77973-670
Counstar Spica M1 Real-Time Live Cell Imaging and Analysis System
Cell Counters and Flow Cytometers Electronic Cell Counters
The Countstar Spica provides real-time live cell imaging and analysis to support a wide variety of applications in life science research, drug development, and cell-based therapies.
  • Incubator-friendly for optimal cell health
  • Monitor experiments from anywhere, anytime for maximu flexibility and efficiency
  • Ready-to-go assays with pre-set parameters for consistent and reliable data for all users
  • Seamless imaging to analysis in one software
  • Unparalleled flexibility for diverse live cell assays
The integration of advanced scanning technology and AI-powered image processing algorithms enables fast and accurate analyses of both 2D cell cultures and 3D organoids. The comprehensive suite of workflow solutions simplifies operation while providing the flexibility to customize and expand the scope of applications to meet new challenges.